4-Hydroxy-3,5-diiodobenzyl alcohol ≥97%

Supplier: Matrix Scientific

114793-1G
102894-714EA 1298.46 USD
102894-714
4-Hydroxy-3,5-diiodobenzyl alcohol ≥97%
4-Hydroxy-3,5-diiodobenzyl alcohol

Hazard: Irritant

Formula: C₇H₆I₂O₂
MDL Number: MFCD06203112
CAS Number: 37987-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR