5-Bromo-3-methyl-3H-pyrimidin-4-one ≥97%

Supplier: Matrix Scientific

114776-1G
102894-678EA 1298.46 USD
102894-678
5-Bromo-3-methyl-3H-pyrimidin-4-one ≥97%
5-Bromo-3-methyl-3H-pyrimidin-4-one

Hazard: Irritant

Formula: C₅H₅BrN₂O
Melting Pt: 152…155 °C
MDL Number: MFCD00834961
CAS Number: 14248-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR