2-Iodo-5-methyl-1,3-thiazole ≥97%

Supplier: Matrix Scientific

114772-1G
102898-912EA 958.46 USD
102898-912
2-Iodo-5-methyl-1,3-thiazole ≥97%
2-Iodo-5-methyl-1,3-thiazole

Hazard: Irritant

Formula: C₄H₄INS
MDL Number: MFCD20484739
CAS Number: 847547-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR