2-Chloro-5-methylthiazole ≥97%

Supplier: Matrix Scientific

114771-1G
102898-910EA 369.23 USD
102898-910
2-Chloro-5-methylthiazole ≥97%
2-Chloro-5-methylthiazole

Hazard: Irritant

Formula: C₄H₄ClNS
MW: 133.6000 g/mol
MDL Number: MFCD03265390
CAS Number: 33342-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR