2-Bromo-5-(methylthio)pyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-5-methylthiopyridine

114767-1G
102898-900EA 1223.08 USD
102898-900
2-Bromo-5-(methylthio)pyridine ≥97%
2-Bromo-5-(methylthio)pyridine

Hazard: Irritant

Formula: C₆H₆BrNS
MW: 204.09 g/mol
MDL Number: MFCD04974522
CAS Number: 134872-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR