2-Bromo-6-phenylpyridine ≥97%

Supplier: Matrix Scientific

114764-1G
102898-894EA 447.69 USD
102898-894
2-Bromo-6-phenylpyridine ≥97%
2-Bromo-6-phenylpyridine

Hazard: Irritant

Formula: C₁₁H₈BrN
MDL Number: MFCD00234691
CAS Number: 39774-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR