6-Bromo-7-methylquinoxaline ≥97%

Supplier: Matrix Scientific

114763-1G
102898-892EA 1284.62 USD
102898-892
6-Bromo-7-methylquinoxaline ≥97%
6-Bromo-7-methylquinoxaline

Hazard: Irritant

Formula: C₉H₇BrN₂
MDL Number: MFCD16987756
CAS Number: 646504-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR