5-Chloro-2,1,3-benzoxadiazole ≥97%

Supplier: Matrix Scientific

114751-1G
102898-466EA 206.15 USD
102898-466
5-Chloro-2,1,3-benzoxadiazole ≥97%
5-Chloro-2,1,3-benzoxadiazole

Hazard: Irritant

Formula: C₆H₃ClN₂O
MDL Number: MFCD00084936
CAS Number: 19155-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR