2,6-Dichloro-1H-benzimidazole ≥97%

Supplier: Matrix Scientific

114750-1G
102898-464EA 813.85 USD
102898-464
2,6-Dichloro-1H-benzimidazole ≥97%
2,6-Dichloro-1H-benzimidazole

Hazard: Irritant

Formula: C₇H₄Cl₂N₂
MDL Number: MFCD11044660
CAS Number: 4887-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR