1-(3-Bromophenyl)-2,2,2-trifluoroethanol ≥97%

Supplier: Matrix Scientific

114745-1G
102894-666EA 958.46 USD
102894-666
1-(3-Bromophenyl)-2,2,2-trifluoroethanol ≥97%
1-(3-Bromophenyl)-2,2,2-trifluoroethanol

Hazard: Irritant

Formula: C₈H₆BrF₃O
MDL Number: MFCD16067907
CAS Number: 446-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR