2-(3-Fluorophenyl)piperazine ≥97%

Supplier: Matrix Scientific

114736-1G
102894-648EA 738.46 USD
102894-648
2-(3-Fluorophenyl)piperazine ≥97%
2-(3-Fluorophenyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₃FN₂
MDL Number: MFCD06411150
CAS Number: 137684-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR