Diethyl-3,3'-thiobispropionate ≥97%

Supplier: Matrix Scientific

114733-1G
102894-640EA 253.85 USD
102894-640
Diethyl-3,3'-thiobispropionate ≥97%
Diethyl-3,3'-thiobispropionate

Hazard: Irritant

Formula: C₁₀H₁₈O₄S
MDL Number: MFCD00026920
CAS Number: 673-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR