Ethyl-4-oxotetrahydro-2H-thiopyran-3-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-oxotetrahydro-2H-thiopyran-3-carboxylate

114726-1G
102894-608EA 958.46 USD
102894-608
Ethyl-4-oxotetrahydro-2H-thiopyran-3-carboxylate ≥97%
Ethyl-4-oxotetrahydro-2H-thiopyran-3-carboxylate

Hazard: Irritant

Formula: C₈H₁₂O₃S
MW: 188.25 g/mol
MDL Number: MFCD00040797
CAS Number: 1198-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR