Methyl-3-methylenecyclobutanecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3-methylenecyclobutanecarboxylate

114723-1G
102894-602EA 261.54 USD
102894-602
Methyl-3-methylenecyclobutanecarboxylate ≥97%
Methyl-3-methylenecyclobutanecarboxylate

Hazard: Irritant

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
MDL Number: MFCD01087180
CAS Number: 15963-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR