Methyl-3-thiomorpholinecarboxylate ≥97%

Supplier: Matrix Scientific

114712-1G
102894-578EA 958.46 USD
102894-578
Methyl-3-thiomorpholinecarboxylate ≥97%
Methyl-3-thiomorpholinecarboxylate

Hazard: Irritant

Formula: C₆H₁₁NO₂S
MDL Number: MFCD09405990
CAS Number: 78865-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR