Ethyl-2-(piperidin-4-yl)propanoate ≥97%

Supplier: Matrix Scientific

114711-1G
102894-576EA 3272.31 USD
102894-576
Ethyl-2-(piperidin-4-yl)propanoate ≥97%
Ethyl-2-(piperidin-4-yl)propanoate

Hazard: Irritant

Formula: C₁₀H₁₉NO₂
MDL Number: MFCD20668512
CAS Number: 141060-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR