Methyl-1-methylpiperidine-4-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 1-methylpiperidine-4-carboxylate
114710-1G
102894-574EA
350.77
USD
102894-574
Methyl-1-methylpiperidine-4-carboxylate ≥97%
Methyl-1-methylpiperidine-4-carboxylate
Hazard: Irritant
Formula:
C₈H₁₅NO₂ MW: 157.21 g/mol |
MDL Number:
MFCD06658483 CAS Number: 1690-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...