Methyl-1-(tert-butoxycarbonyl)-piperidine-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 1-Boc-2-piperidinecarboxylateMethyl 1-(tert-Butoxycarbonyl)-2-piperidinecarboxylate1-tert-Butyl 2-Methyl Piperidine-1,2-dicarboxylatePiperidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl ester1-(tert-Butoxycarbonyl)-2-piperidinecarboxylic acid methyll ester1-Boc-2-piperidinecarboxylic acid methyll ester

114707-1G
102894-568EA 690.77 USD
102894-568
Methyl-1-(tert-butoxycarbonyl)-piperidine-2-carboxylate ≥97%
Methyl-1-(tert-butoxycarbonyl)-piperidine-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.31 g/mol
MDL Number: MFCD06795925
CAS Number: 167423-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR