Ethyl-4-aminopyrimidine-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-aminopyrimidine-5-carboxylate

114694-1G
102894-542EA 738.46 USD
102894-542
Ethyl-4-aminopyrimidine-5-carboxylate ≥97%
Ethyl-4-aminopyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₇H₉N₃O₂
MW: 167.17 g/mol
MDL Number: MFCD01028678
CAS Number: 65195-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR