Ethyl-4-ethoxy-2-(methylthio)pyrimidine-5-carboxylate ≥97%

Supplier: Matrix Scientific

114690-1G
102894-534EA 813.85 USD
102894-534
Ethyl-4-ethoxy-2-(methylthio)pyrimidine-5-carboxylate ≥97%
Ethyl-4-ethoxy-2-(methylthio)pyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₃S
MDL Number: MFCD16660585
CAS Number: 84923-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR