Ethyl-4-cyclopropyl-1,3-thiazole-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-cyclopropylthiazole-2-carboxylate

114681-1G
102894-498EA 544.62 USD
102894-498
Ethyl-4-cyclopropyl-1,3-thiazole-2-carboxylate ≥97%
Ethyl-4-cyclopropyl-1,3-thiazole-2-carboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₂S
MW: 197.26 g/mol
MDL Number: MFCD11656780
CAS Number: 439692-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR