Ethyl-2-(methoxymethyl)-4-methyl-1,3-thiazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

114680-1G
102894-496EA 1104.62 USD
102894-496
Ethyl-2-(methoxymethyl)-4-methyl-1,3-thiazole-5-carboxylate ≥97%
Ethyl-2-(methoxymethyl)-4-methyl-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₉H₁₃NO₃S
MDL Number: MFCD28099757
CAS Number: 136204-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR