Ethyl-2-(methoxymethyl)-1,3-thiazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

114679-1G
102894-494EA 958.46 USD
102894-494
Ethyl-2-(methoxymethyl)-1,3-thiazole-4-carboxylate ≥97%
Ethyl-2-(methoxymethyl)-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₈H₁₁NO₃S
MDL Number: MFCD17018002
CAS Number: 854754-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR