Dimethyl-2-(1H-pyrrol-1-yl)pentanedioate ≥97%

Supplier: Matrix Scientific

114675-1G
102894-486EA 958.46 USD
102894-486
Dimethyl-2-(1H-pyrrol-1-yl)pentanedioate ≥97%
Dimethyl-2-(1H-pyrrol-1-yl)pentanedioate

Hazard: Irritant

Formula: C₁₁H₁₅NO₄
MDL Number: MFCD00728861
CAS Number: 259655-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR