Ethyl-5-[(acetyloxy)methyl]-2-furoate ≥97%

Supplier: Matrix Scientific

114674-1G
102894-484EA 570.77 USD
102894-484
Ethyl-5-[(acetyloxy)methyl]-2-furoate ≥97%
Ethyl-5-[(acetyloxy)methyl]-2-furoate

Hazard: Irritant

Formula: C₁₀H₁₂O₅
MDL Number: MFCD00068175
CAS Number: 99187-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR