Ethyl-3-oxo-4-(thiophene-2-yl)butanoate ≥97%
Supplier: Matrix Scientific
114668-1G
102894-468EA
958.46
USD
102894-468
Ethyl-3-oxo-4-(thiophene-2-yl)butanoate ≥97%
Ethyl-3-oxo-4-(thiophene-2-yl)butanoate
Hazard: Irritant
Formula:
C₁₀H₁₂O₃S |
MDL Number:
MFCD16779559 CAS Number: 86728-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...