Methyl-3-nitroisonicotinoate ≥97%

Supplier: Matrix Scientific

114663-1G
102894-458EA 738.46 USD
102894-458
Methyl-3-nitroisonicotinoate ≥97%
Methyl-3-nitroisonicotinoate

Hazard: Irritant

Formula: C₇H₆N₂O₄
MDL Number: MFCD16657194
CAS Number: 103698-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR