Methyl-3,5-dimethoxy-4-methylbenzoate ≥97%

Supplier: Matrix Scientific

114644-1G
102894-418EA 958.46 USD
102894-418
Methyl-3,5-dimethoxy-4-methylbenzoate ≥97%
Methyl-3,5-dimethoxy-4-methylbenzoate

Hazard: Irritant

Formula: C₁₁H₁₄O₄
MDL Number: MFCD00017512
CAS Number: 60441-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR