Methyl-2-iodo-4-methoxybenzoate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-iodo-4-methoxybenzoate
114636-1G
102894-402EA
958.46
USD
102894-402
Methyl-2-iodo-4-methoxybenzoate ≥97%
Methyl-2-iodo-4-methoxybenzoate
Hazard: Irritant
Formula:
C₉H₉IO₃ MW: 292.07 g/mol |
MDL Number:
MFCD18905183 CAS Number: 54413-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...