Methyl-2-(4-fluorophenyl)-2-hydroxyacetate ≥97%
Supplier: Matrix Scientific
114622-1G
102898-440EA
813.85
USD
102898-440
Methyl-2-(4-fluorophenyl)-2-hydroxyacetate ≥97%
Methyl-2-(4-fluorophenyl)-2-hydroxyacetate
Hazard: Irritant
Formula:
C₉H₉FO₃ MW: 184.17 g/mol |
MDL Number:
MFCD16681137 CAS Number: 127709-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...