3,3,3-Trifluoro-2,2-dimethylpropanoyl chloride ≥95%

Supplier: Matrix Scientific

114617-1G
102898-430EA 958.46 USD
102898-430
3,3,3-Trifluoro-2,2-dimethylpropanoyl chloride ≥95%
3,3,3-Trifluoro-2,2-dimethylpropanoyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₅H₆ClF₃O
MDL Number: MFCD22199505
CAS Number: 1163707-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR