(2E)-Pent-2-enoyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: (E)-Pent-2-enoyl chloride

114615-1G
102898-426EA 813.85 USD
102898-426
(2E)-Pent-2-enoyl chloride ≥95%
(2E)-Pent-2-enoyl chloride

Hazard: Irritant

Formula: C₅H₇ClO
MW: 118.56 g/mol
MDL Number: MFCD00040130
CAS Number: 33698-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR