(7-Methoxy-1-naphthyl)acetyl chloride ≥97%
Supplier: Matrix Scientific
114606-1G
102894-384EA
958.46
USD
102894-384
(7-Methoxy-1-naphthyl)acetyl chloride ≥97%
(7-Methoxy-1-naphthyl)acetyl chloride
Hazard: Irritant
Formula:
C₁₃H₁₁ClO₂ MW: 234.68 g/mol |
MDL Number:
MFCD27956331 CAS Number: 6836-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...