2-Methylimidazo[1,2-a]pyridine-3-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114603-1G
102894-378EA 958.46 USD
102894-378
2-Methylimidazo[1,2-a]pyridine-3-carbonyl chloride ≥97%
2-Methylimidazo[1,2-a]pyridine-3-carbonyl chloride

Hazard: Irritant

Formula: C₉H₇ClN₂O
MDL Number: MFCD18459043
CAS Number: 135854-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR