1-Methyl-1H-indazole-3-carbonyl chloride ≥97%
Supplier: Matrix Scientific
114598-1G
102894-368EA
315.38
USD
102894-368
1-Methyl-1H-indazole-3-carbonyl chloride ≥97%
1-Methyl-1H-indazole-3-carbonyl chloride
Hazard: Irritant
Formula:
C₉H₇ClN₂O |
MDL Number:
MFCD02093096 CAS Number: 106649-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...