2-(2-Oxocyclohexyl)acetyl chloride ≥97%

Supplier: Matrix Scientific

114593-1G
102894-358EA 1443.08 USD
102894-358
2-(2-Oxocyclohexyl)acetyl chloride ≥97%
2-(2-Oxocyclohexyl)acetyl chloride

Hazard: Irritant

Formula: C₈H₁₁ClO₂
MDL Number: MFCD24693274
CAS Number: 342402-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR