4-(Aminomethyl)cyclohexanecarbonyl chloride ≥97%

Supplier: Matrix Scientific

114589-1G
102894-350EA 958.46 USD
102894-350
4-(Aminomethyl)cyclohexanecarbonyl chloride ≥97%
4-(Aminomethyl)cyclohexanecarbonyl chloride

Hazard: Irritant

Formula: C₈H₁₄ClNO
MDL Number: MFCD24507119
CAS Number: 78044-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR