Cycloheptanecarbonyl chloride ≥97%

Supplier: Matrix Scientific

114586-1G
102894-344EA 884.62 USD
102894-344
Cycloheptanecarbonyl chloride ≥97%
Cycloheptanecarbonyl chloride

Hazard: Irritant

Formula: C₈H₁₃ClO
MDL Number: MFCD11520758
CAS Number: 6557-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR