5-Bromo-2,3-dihydrobenzo[b]furan-7-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114580-1G
102894-332EA 958.46 USD
102894-332
5-Bromo-2,3-dihydrobenzo[b]furan-7-carbonyl chloride ≥97%
5-Bromo-2,3-dihydrobenzo[b]furan-7-carbonyl chloride

Hazard: Irritant

Formula: C₉H₆BrClO₂
MW: 261.51 g/mol
MDL Number: MFCD05865150
CAS Number: 108551-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR