2,3-Dihydro-1-benzofuran-7-carbonyl chloride ≥95%
Supplier: Matrix Scientific
114579-1G
102894-330EA
958.46
USD
102894-330
2,3-Dihydro-1-benzofuran-7-carbonyl chloride ≥95%
2,3-Dihydro-1-benzofuran-7-carbonyl chloride
Hazard: Irritant
Formula:
C₉H₇ClO₂ MW: 182.61 g/mol |
MDL Number:
MFCD06658967 CAS Number: 123266-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...