Quinoline-4-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114577-1G
102894-326EA 813.85 USD
102894-326
Quinoline-4-carbonyl chloride ≥97%
Quinoline-4-carbonyl chloride

Hazard: Irritant

Formula: C₁₀H₆ClNO
MDL Number: MFCD15523600
CAS Number: 50821-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR