Quinoline-6-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114576-1G
102894-324EA 884.62 USD
102894-324
Quinoline-6-carbonyl chloride ≥97%
Quinoline-6-carbonyl chloride

Hazard: Irritant

Formula: C₁₀H₆ClNO
MDL Number: MFCD15523598
CAS Number: 72369-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR