1,2,3-Benzothiadiazole-5-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114575-1G
102894-322EA 716.92 USD
102894-322
1,2,3-Benzothiadiazole-5-carbonyl chloride ≥97%
1,2,3-Benzothiadiazole-5-carbonyl chloride

Hazard: Irritant

Formula: C₇H₃ClN₂OS
MDL Number: MFCD01764004
CAS Number: 321309-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR