1,3-Benzothiazole-6-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114574-1G
102894-320EA 884.62 USD
102894-320
1,3-Benzothiazole-6-carbonyl chloride ≥97%
1,3-Benzothiazole-6-carbonyl chloride

Hazard: Irritant

Formula: C₈H₄ClNOS
MDL Number: MFCD04972624
CAS Number: 55439-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR