5-Bromo-1-benzothiophene-3-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114564-1G
102894-300EA 641.54 USD
102894-300
5-Bromo-1-benzothiophene-3-carbonyl chloride ≥97%
5-Bromo-1-benzothiophene-3-carbonyl chloride

Hazard: Irritant

Formula: C₉H₄BrClOS
MDL Number: MFCD07368522
CAS Number: 850374-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR