6-Chloropyridazine-3-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114561-1G
102894-294EA 958.46 USD
102894-294
6-Chloropyridazine-3-carbonyl chloride ≥97%
6-Chloropyridazine-3-carbonyl chloride

Hazard: Irritant

Formula: C₅H₂Cl₂N₂O
MDL Number: MFCD09991889
CAS Number: 6531-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR