1H-Pyrazole-4-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114553-1G
102898-410EA 958.46 USD
102898-410
1H-Pyrazole-4-carbonyl chloride ≥97%
1H-Pyrazole-4-carbonyl chloride

Hazard: Irritant

Formula: C₄H₃ClN₂O
MDL Number: MFCD17289359
CAS Number: 103286-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR