1-Methylindoline-5-carboxylic acid ≥97%

Supplier: Matrix Scientific

114539-1G
102894-252EA 884.62 USD
102894-252
1-Methylindoline-5-carboxylic acid ≥97%
1-Methylindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD09879911
CAS Number: 380922-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR