2-Methyl-1,3-thiazole-5-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114511-1G
102894-196EA 1272.31 USD
102894-196
2-Methyl-1,3-thiazole-5-carbonyl chloride ≥97%
2-Methyl-1,3-thiazole-5-carbonyl chloride

Hazard: Irritant

Formula: C₅H₄ClNOS
MDL Number: MFCD13173719
CAS Number: 60971-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR