3-(Trifluoromethyl)pyridine-2-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114505-1G
102894-182EA 884.62 USD
102894-182
3-(Trifluoromethyl)pyridine-2-carbonyl chloride ≥97%
3-(Trifluoromethyl)pyridine-2-carbonyl chloride

Hazard: Irritant

Formula: C₇H₃ClF₃NO
MDL Number: MFCD13174647
CAS Number: 128073-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR